Tisk…

2-Cyclohexylbenzoic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

L18060.03 L18060.06
L18060.03EA 2830 CZK
L18060.03 L18060.06
2-Cyclohexylbenzoic acid 97%
2-Cyclohexylbenzoic acid

Purity: 97% CAS No.: 97023-48-8 Molecular Formula: C13H16O2 Formula Weight: 204.27 Melting Point: 103-105° Beilstein Reference: 2452158 MDL No.: MFCD00452653

Vzorec: C₁₃H₁₆O₂
Bod tání: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00452653
CAS číslo: 97023-48-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR