5-(4-Fluorophenyl)valeric acid 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
5-(4-Fluorophenyl)pentanoic acid
L17935.03
L17935.06
L17935.MD
L17935.03EA
2970
CZK
L17935.03
L17935.06
L17935.MD
5-(4-Fluorophenyl)valeric acid 97%
5-(4-Fluorophenyl)valeric acid
Purity: 97%
CAS No.: 24484-22-8
Molecular Formula: C11H13FO2
Formula Weight: 196.22
Melting Point: 73-75°
Beilstein Reference: 7918899
MDL No.: MFCD00800242
Vzorec:
C₁₁H₁₃FO₂ MW: 196,22 g/mol Bod tání: 73…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00800242 CAS číslo: 24484-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...