7-Bromo-1-hydroxyisoquinoline ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
L17800.06
L17800.03
L17800.06EA
9960
CZK
L17800.06
L17800.03
7-Bromo-1-hydroxyisoquinoline ≥98%
7-Bromo-1-hydroxyisoquinoline
Purity: 98+%
CAS No.: 223671-15-6
Molecular Formula: C9H6BrNO
Formula Weight: 224.06
Melting Point: 248-250°
Beilstein Reference: 8405244
MDL No.: MFCD02093963
Vzorec:
C₉H₆BrNO MW: 224,06 g/mol Bod tání: 248…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093963 CAS číslo: 223671-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...