Tisk…

1-(Phenylsulphonyl)indole 98%

Dodavatel: Thermo Scientific

Synonyms: 1-(Phenylsulphonyl)-1H-indole

L17566.03 L17566.06
L17566.03EA 766 CZK
L17566.03 L17566.06
1-(Phenylsulphonyl)indole 98%
1-(Phenylsulphonyl)indole

Purity: 98% CAS No.: 40899-71-6 Molecular Formula: C14H11NO2S Formula Weight: 257.31 Melting Point: 77-79° Boiling Point: 192°/3mm Beilstein Reference: 1286712 MDL No.: MFCD00134318

Vzorec: C₁₄H₁₁NO₂S
MW: 257,31 g/mol
Bod varu: 192 °C (3 mmHg)
Bod tání: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00134318
CAS číslo: 40899-71-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR