1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L17528.06
L17528.03
L17528.06EA
1690
CZK
L17528.06
L17528.03
1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid 98%
1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
Purity: 98%
CAS No.: 98303-20-9
Molecular Formula: C11H19NO4
Formula Weight: 229.28
Melting Point: 129-131°
Beilstein Reference: 480022
MDL No.: MFCD01862877
Vzorec:
C₁₁H₁₉NO₄ Bod tání: 129…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD01862877 CAS číslo: 98303-20-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...