Ethyl-2-methyl-4,4,4-trifluoroacetoacetate 96%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Ethyl 4,4,4-Trifluoro-2-methyl-3-oxobutyrate
L17335.03
L17335.06
L17335.03EA
1010
CZK
L17335.03
L17335.06
Ethyl-2-methyl-4,4,4-trifluoroacetoacetate 96%
Ethyl-2-methyl-4,4,4-trifluoroacetoacetate
Purity: 96%
CAS No.: 344-00-3
Molecular Formula: C7H9F3O3
Formula Weight: 198.14
Flash Point: 47°(116°F)
Refractive Index: 1.3700
Boiling Point: 144-146°
Beilstein Reference: 1783620
UN No.: UN3272
MDL No.: MFCD00190635
Vzorec:
C₇H₉F₃O₃ MW: 198,14 g/mol Bod varu: 144…146 °C Hustota: 1,258 Bod vzplanutí: 47 °C (116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190635 CAS číslo: 344-00-3 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...