Ethyl-5-methylindole-2-carboxylate 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Ethyl 5-methylindole-2-carboxylate
L17146.03
L17146.06
L17146.03EA
1220
CZK
L17146.03
L17146.06
Ethyl-5-methylindole-2-carboxylate 98%
Ethyl-5-methylindole-2-carboxylate
Purity: 99%
CAS No.: 16382-15-3
Molecular Formula: C12H13NO2
Formula Weight: 203.24
Melting Point: 161-165°
Boiling Point: 236°/4mm
Beilstein Reference: 159437
MDL No.: MFCD00022703
Vzorec:
C₁₂H₁₃NO₂ Bod varu: 236 °C (4 mmHg) Bod tání: 161…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022703 CAS číslo: 16382-15-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...