6-Cyanonicotinic acid 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L17127.03
L17127.MD
L17127.03EA
5840
CZK
L17127.03
L17127.MD
6-Cyanonicotinic acid 97%
6-Cyanonicotinic acid
Purity: 97%
CAS No.: 70165-31-0
Molecular Formula: C7H4N2O2
Formula Weight: 148.12
Melting Point: 189-194°
Beilstein Reference: 4384027
MDL No.: MFCD01318135
Vzorec:
C₇H₄N₂O₂ MW: 148,12 g/mol Bod tání: 189…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318135 CAS číslo: 70165-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...