Tisk…

N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1-(Chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane Bis(tetrafluoroborate), F-TEDA-BF4, 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)

L17003.06 L17003.14
L17003.06EA 1860 CZK
L17003.06 L17003.14
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥98%
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)

Purity: 98+% CAS No.: 140681-55-6 Molecular Formula: C7H14B2ClF9N2 Formula Weight: 354.26 Melting Point: ca 250° dec. UN No.: UN3088 MDL No.: MFCD00142607

Vzorec: C₇H₁₄B₂ClF₉N₂
MW: 354,26 g/mol
Bod tání: ∼ 250 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00142607
CAS číslo: 140681-55-6
EINECS: 414-380-4
ADR: 4.2,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR