Tisk…

2-Methoxy-4-methylbenzenesulphonamide 95%

Dodavatel: Thermo Scientific

L16947.03 L16947.06
L16947.03EA 851 CZK
L16947.03 L16947.06
2-Methoxy-4-methylbenzenesulphonamide 95%
2-Methoxy-4-methylbenzenesulphonamide

Purity: 95% CAS No.: 59554-39-1 Molecular Formula: C8H11NO3S Formula Weight: 201.25 Melting Point: 168-170° MDL No.: MFCD01318156

Vzorec: C₈H₁₁NO₃S
Bod tání: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD01318156
CAS číslo: 59554-39-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR