Tisk…

Trimethoxy[3-(phenylamino)propyl]silane 96%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L16850.14 L16850.22
L16850.14EA 999 CZK
L16850.14 L16850.22
Trimethoxy[3-(phenylamino)propyl]silane 96%
Trimethoxy[3-(phenylamino)propyl]silane

Purity: 96% CAS No.: 3068-76-6 Molecular Formula: C12H21NO3Si Formula Weight: 255.39 Flash Point: 165°(329°F) Refractive Index: 1.5055 Odour: Weak Boiling Point: 310° Beilstein Reference: 8620045 UN No.: UN3267 MDL No.: MFCD00053673

Vzorec: C₁₂H₂₁NO₃Si
Bod varu: 310 °C
Hustota: 1,07
Bod vzplanutí: 165 °C (329 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053673
CAS číslo: 3068-76-6
EINECS: 221-328-2
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR