Tisk…

Dimethylethoxyvinylsilane 97%

Dodavatel: Thermo Scientific
Nebezpečí

L16699.14 L16699.22
L16699.14EA 3710 CZK
L16699.14 L16699.22
Dimethylethoxyvinylsilane 97%
Dimethylethoxyvinylsilane

Purity: 97% CAS No.: 5356-83-2 Molecular Formula: C6H14OSi Formula Weight: 130.26 Flash Point: -3°(26°F) Refractive Index: 1.3980 Boiling Point: 99° Beilstein Reference: 1743477 UN No.: UN1993 MDL No.: MFCD00009070

Vzorec: C₆H₁₄OSi
Bod varu: 99 °C
Hustota: 0,790
Bod vzplanutí: –3 °C (26 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009070
CAS číslo: 5356-83-2
EINECS: 226-341-7
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR