Tisk…

2-(Trimethylsilyloxy)ethyl methacrylate 94% stabilizovaný

Dodavatel: Thermo Scientific
Varování

Synonyms: Methacrylic Acid 2-(Trimethylsilyloxy)ethyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 100 ppm

L16675.18 L16675.09
L16675.18EA 3540 CZK
L16675.18 L16675.09
2-(Trimethylsilyloxy)ethyl methacrylate 94% stabilizovaný
2-(Trimethylsilyloxy)ethyl methacrylate

Stab. with 100ppm 4-methoxyphenol


Purity: 94% CAS No.: 17407-09-9 Molecular Formula: C9H18O3Si Formula Weight: 202.33 Flash Point: 42°(107°F) Refractive Index: 1.4300 Boiling Point: 65°/0.9mm MDL No.: MFCD00053869

Vzorec: C₉H₁₈O₃Si
MW: 202,33 g/mol
Bod varu: 65 °C (0,9 mmHg)
Hustota: 0,995
Bod vzplanutí: 42 °C (107 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053869
CAS číslo: 17407-09-9
EINECS: 241-432-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR