Tisk…

Ethoxytriethylsilane 97%

Dodavatel: Thermo Scientific
Varování

L16656.06 L16656.14
L16656.06EA 1920 CZK
L16656.06 L16656.14
Ethoxytriethylsilane 97%
Ethoxytriethylsilane

Purity: 97% CAS No.: 597-67-1 Molecular Formula: C8H20OSi Formula Weight: 160.33 Refractive Index: 1.4140 Boiling Point: 154-155° Beilstein Reference: 1735027 UN No.: UN1993 MDL No.: MFCD00054718

Vzorec: C₈H₂₀OSi
Bod varu: 154…155 °C
Hustota: 0,816
Storage Temperature: Ambient
MDL Number: MFCD00054718
CAS číslo: 597-67-1
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR