Tisk…

Undecafluoro(2-methyl-3-oxahexanoyl)fluoride 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Perfluoro(2-methyl-3-oxahexanoyl) fluoride

L16483.06 L16483.14
L16483.06EA 1950 CZK
L16483.06 L16483.14
Undecafluoro(2-methyl-3-oxahexanoyl)fluoride 97%
Undecafluoro(2-methyl-3-oxahexanoyl)fluoride

Purity: 97% CAS No.: 2062-98-8 Molecular Formula: C6F12O2 Formula Weight: 332.06 Flash Point: None Refractive Index: 1.3000 Boiling Point: 54-56° UN No.: UN3265 MDL No.: MFCD00054657

Vzorec: C₆F₁₂O₂
MW: 332.05 g/mol
Bod varu: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00054657
CAS číslo: 2062-98-8
EINECS: 218-173-8
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR