Tisk…

Trimorpholinophosphine oxide 99%

Dodavatel: Thermo Fisher Scientific
Varování

L16315.06 L16315.14
L16315.06EA 1260 CZK
L16315.06 L16315.14
Trimorpholinophosphine oxide 99%
Trimorpholinophosphine oxide

Purity: 99% CAS No.: 4441-12-7 Molecular Formula: C12H24N3O4P Formula Weight: 305.32 Melting Point: 188-192° Beilstein Reference: 280155 MDL No.: MFCD00047406

Vzorec: C₁₂H₂₄N₃O₄P
Bod tání: 188…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00047406
CAS číslo: 4441-12-7
EINECS: 224-662-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR