Tisk…

4,4'-Oxybis(benzoic acid) ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4,4′-Oxybis(benzoic acid)

L15651.14 L15651.22
L15651.14EA 1340 CZK
L15651.14 L15651.22
4,4'-Oxybis(benzoic acid) ≥98%
4,4'-Oxybis(benzoic acid)

Purity: 98+% CAS No.: 2215-89-6 Molecular Formula: C14H10O5 Formula Weight: 258.23 Melting Point: 329-332° Beilstein Reference: 2218315 MDL No.: MFCD00013988

Vzorec: C₁₄H₁₀O₅
Bod tání: 329…332 °C
Storage Temperature: Ambient
MDL Number: MFCD00013988
CAS číslo: 2215-89-6
EINECS: 218-683-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR