5-Methoxyindole-2-carboxylic acid 97%
Dodavatel: Thermo Fisher Scientific
L15137.03
L15137.06
L15137.03EA
790
CZK
L15137.03
L15137.06
5-Methoxyindole-2-carboxylic acid 97%
5-Methoxyindole-2-carboxylic acid
Purity: 97%
CAS No.: 4382-54-1
Molecular Formula: C10H9NO3
Formula Weight: 191.19
Melting Point: ca 200° dec.
Beilstein Reference: 157478
MDL No.: MFCD00005614
Vzorec:
C₁₀H₉NO₃ MW: 191.19 g/mol Bod tání: ∼ 200 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00005614 CAS číslo: 4382-54-1 EINECS: 224-487-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...