Benzimidazole-5,6-dicarboxylic acid 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1H-Benzo[d]imidazole-5,6-dicarboxylic acid
L14754.06
L14754.03
L14754.06EA
5460
CZK
L14754.06
L14754.03
Benzimidazole-5,6-dicarboxylic acid 97%
Benzimidazole-5,6-dicarboxylic acid
Purity: 97%
CAS No.: 10351-75-4
Molecular Formula: C9H6N2O4
Formula Weight: 206.16
Melting Point: ca 295° dec.
Beilstein Reference: 528170
MDL No.: MFCD03093058
Vzorec:
C₉H₆N₂O₄ MW: 206,16 g/mol Bod tání: ∼ 295 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD03093058 CAS číslo: 10351-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...