Tisk…

3,4,9,10-Perylenetetracarboxylic dianhydride 98%

Dodavatel: Thermo Scientific
Nebezpečí

L14370.09 L14370.18
L14370.09EA 803 CZK
L14370.09 L14370.18
3,4,9,10-Perylenetetracarboxylic dianhydride 98%
3,4,9,10-Perylenetetracarboxylic dianhydride

Purity: 98% CAS No.: 128-69-8 Molecular Formula: C24H8O6 Formula Weight: 392.32 Odour: Odourless Melting Point: 300° Beilstein Reference: 57831 MDL No.: MFCD00006916

Vzorec: C₂₄H₈O₆
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006916
CAS číslo: 128-69-8
EINECS: 204-905-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR