Naphthalene-1,4,5,8-tetracarboxylic dianhydride 97%
Dodavatel: Thermo Scientific
|
Varování
|
L14365.06
L14365.14
L14365.06EA
1480
CZK
L14365.06
L14365.14
Naphthalene-1,4,5,8-tetracarboxylic dianhydride 97%
Naphthalene-1,4,5,8-tetracarboxylic dianhydride
Purity: 97%
CAS No.: 81-30-1
Molecular Formula: C14H4O6
Formula Weight: 268.18
Melting Point:
Vzorec:
C₁₄H₄O₆ Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006915 CAS číslo: 81-30-1 EINECS: 201-342-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...