Lauroyl peroxide 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Dilauroyl peroxide
L14310.22
L14310.36
L14310.22EA
1640
CZK
L14310.22
L14310.36
Lauroyl peroxide 97%
Lauroyl peroxide
Purity: 97%
CAS No.: 105-74-8
Molecular Formula: [CH3(CH2)10CO2]2
Formula Weight: 398.63
Melting Point: 53-57°
Beilstein Reference: 1804936
UN No.: UN3106
MDL No.: MFCD00008964
Vzorec:
(C₁₁H₂₃CO)₂O₂ Bod tání: 53…57 °C Hustota: 0,910 Storage Temperature: Ambient |
MDL Number:
MFCD00008964 CAS číslo: 105-74-8 EINECS: 203-326-3 ADR: 5.2,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...