Tisk…
Dowex® 1X2 (Cl) 100-200 mesh
Dodavatel: Thermo Fisher Scientific
L14254.18 L14254.30 L14254.0B
L14254.18EA 1590 CZK
L14254.18 L14254.30 L14254.0B
Dowex® 1X2 (Cl) 100-200 mesh
Dowex® 1X2 (Cl)
CAS No.: 9085-42-1 Odour: Amine-like MDL No.: MFCD00132715
Storage Temperature: Ambient
MDL Number: MFCD00132715
CAS číslo: 9085-42-1