S-(2-Aminoethyl)isothiouronium bromide hydrobromide 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
S-(2-Aminoethyl)isothiourea Dihydrobromide, 2-(2-Aminoethyl)-2-thiopseudourea Dihydrobromide, S-(2-Aminoethyl)thiouronium bromide hydrobromide, AET, Carbamimidothioic Acid 2-Aminoethyl Ester Dihydrobromide
Purity: 98%
CAS No.: 56-10-0
Molecular Formula: C3H9N3S.2HBr
Formula Weight: 281.02
Odour: Odourless
Melting Point: 190-192°
Beilstein Reference: 3911163
MDL No.: MFCD00037011
Vzorec:
C₃H₁₁Br₂N₃S MW: 281,01 g/mol Bod tání: 190…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037011 CAS číslo: 56-10-0 EINECS: 200-257-0 Merck Index: 14,178 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...