Tisk…

4-Carbethoxyhexafluorobutyryl chloride 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Ethyl hexafluoroglutaryl chloride

L14049.03 L14049.06
L14049.03EA 3040 CZK
L14049.03 L14049.06
4-Carbethoxyhexafluorobutyryl chloride 97%
4-Carbethoxyhexafluorobutyryl chloride

Purity: 97% CAS No.: 18381-53-8 Molecular Formula: C7H5ClF6O3 Formula Weight: 286.56 Refractive Index: 1.3590 Boiling Point: 158-160° Beilstein Reference: 1884844 UN No.: UN3265 MDL No.: MFCD00054671

Vzorec: C₇H₅ClF₆O₃
Bod varu: 158…160 °C
Hustota: 1,470
Storage Temperature: Ambient
MDL Number: MFCD00054671
CAS číslo: 18381-53-8
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR