Tisk…

Sodium-3-chloro-2-hydroxypropanesulphonate hemihydrate 98%

Dodavatel: Thermo Scientific
Varování

L14018.36 L14018.22
L14018.36EA 3610 CZK
L14018.36 L14018.22
Sodium-3-chloro-2-hydroxypropanesulphonate hemihydrate 98%
Sodium-3-chloro-2-hydroxypropanesulphonate hemihydrate

Purity: 98% CAS No.: 143218-48-8 Molecular Formula: C3H6ClNaO4S.0.5H2O Formula Weight: 205.60 (196.59anhy) Melting Point: ca 256° dec. Beilstein Reference: 4164376 MDL No.: MFCD00149699

Vzorec: C₃H₆ClNaO₄S·½H₂O
MW: 205,60 g/mol (196,59 anhy)
Bod tání: ∼ 256 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00149699
CAS číslo: 143218-48-8
EINECS: 204-807-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR