N-(Ethoxycarbonyl)nortropinone 99%
Dodavatel: Thermo Scientific
Synonyms:
Ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate, 8-(Ethoxycarbonyl)-8-azabicyclo[3.2.1]octan-3-one, N-Carbethoxy-4-tropinone
L13849.06
L13849.14
L13849.06EA
1670
CZK
L13849.06
L13849.14
N-(Ethoxycarbonyl)nortropinone 99%
N-(Ethoxycarbonyl)nortropinone
Purity: 99%
CAS No.: 32499-64-2
Molecular Formula: C10H15NO3
Formula Weight: 197.23
Refractive Index: 1.4895
Boiling Point: 152-153°/11mm
Beilstein Reference: 1530944
MDL No.: MFCD00078171
Vzorec:
C₁₀H₁₅NO₃ MW: 197,23 g/mol Bod varu: 152…153 °C (11 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00078171 CAS číslo: 32499-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...