8-Methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid 98%
Dodavatel: Thermo Scientific
|
Varování
|
L13751.03
L13751.MD
L13751.03EA
3200
CZK
L13751.03
L13751.MD
8-Methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid 98%
8-Methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid
Purity: 98%
CAS No.: 32178-63-5
Molecular Formula: C12H14O3
Formula Weight: 206.24
Melting Point: 141-143°
Beilstein Reference: 2269890
MDL No.: MFCD01075657
Vzorec:
C₁₂H₁₄O₃ MW: 206,24 g/mol Bod tání: 141…143 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075657 CAS číslo: 32178-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...