Tisk…

Cinnamyl anthranilate 99%

Dodavatel: Thermo Fisher Scientific

L13659.06 L13659.14
L13659.06EA 1850 CZK
L13659.06 L13659.14
Cinnamyl anthranilate 99%
Cinnamyl anthranilate

Purity: 99% CAS No.: 87-29-6 Molecular Formula: C16H15NO2 Formula Weight: 253.30 Melting Point: 64-66° Boiling Point: 322° Beilstein Reference: 2733435 MDL No.: MFCD00053631

Vzorec: C₁₆H₁₅NO₂
Bod varu: 322 °C
Bod tání: 64…66 °C
Hustota: 1,180
Storage Temperature: Ambient
MDL Number: MFCD00053631
CAS číslo: 87-29-6
EINECS: 201-738-8
Merck Index: 14,2303

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR