Methyl-2-methoxypropionate 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Methyl 2-methoxypropanoate
L13527.14
L13527.06
L13527.14EA
5980
CZK
L13527.14
L13527.06
Methyl-2-methoxypropionate 98%
Methyl-2-methoxypropionate
Purity: 98%
CAS No.: 17639-76-8
Molecular Formula: C5H10O3
Formula Weight: 118.13
Flash Point: 36°(96°F)
Refractive Index: 1.3970
Boiling Point: 43-44°/15mm
Beilstein Reference: 1720819
UN No.: UN3272
MDL No.: MFCD00151664
Vzorec:
C₅H₁₀O₃ MW: 118,13 g/mol Bod varu: 43…44 °C (15 mmHg) Bod vzplanutí: 36 °C (96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00151664 CAS číslo: 17639-76-8 EINECS: 241-622-4 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...