trans-3-(4-Pyridyl)acrylic acid 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(E)-3-(Pyridin-4-yl)acrylic acid
L13458.03
L13458.06
L13458.03EA
1700
CZK
L13458.03
L13458.06
trans-3-(4-Pyridyl)acrylic acid 97%
trans-3-(4-Pyridyl)acrylic acid
Purity: 97%
CAS No.: 84228-93-3
Molecular Formula: C8H7NO2
Formula Weight: 149.15
Melting Point: 276-280°
Beilstein Reference: 1719967
MDL No.: MFCD00040746
Vzorec:
C₈H₇NO₂ MW: 149,15 g/mol Bod tání: 276…280 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040746 CAS číslo: 84228-93-3 EINECS: 226-265-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...