Ammeline 90%, tech.
Dodavatel: Thermo Scientific
Synonyms:
Cyanurodiamide, 4,6-Diamino-1,3,5-triazin-2-ol, 2,4-Diamino-6-hydroxy-1,3,5-triazine, 3,4,5,6-Tetrahydro-4,6-diimino-1,3,5-triazin-2(1H)-one
L13064.03
L13064.06
L13064.03EA
1560
CZK
L13064.03
L13064.06
Ammeline 90%, tech.
Ammeline
Purity: 90%
CAS No.: 645-92-1
Molecular Formula: C3H5N5O
Formula Weight: 127.11
Beilstein Reference: 5570
MDL No.: MFCD00023186
Vzorec:
C₃H₅N₅O MW: 127,11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00023186 CAS číslo: 645-92-1 EINECS: 211-455-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...