4-Acryloylmorpholine ≥98% stabilizovaný
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
L12947.06
L12947.14
L12947.06EA
1210
CZK
L12947.06
L12947.14
4-Acryloylmorpholine ≥98% stabilizovaný
4-Acryloylmorpholine
Stab. with 4-methoxyphenol
Purity: 98+%
CAS No.: 5117-12-4
Molecular Formula: C7H11NO2
Formula Weight: 141.17
Storage Temperature: 2°C to 8°C
Flash Point: 127°(260°F)
Refractive Index: 1.5090
Melting Point: -8°
Boiling Point: 158°/50mm
Beilstein Reference: 119302
MDL No.: MFCD00047413
Vzorec:
C₇H₁₁NO₂ MW: 141,17 g/mol Bod varu: 158 °C (50 mmHg) Bod tání: –8 °C Hustota: 1,120 Bod vzplanutí: 127 °C (260 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00047413 CAS číslo: 5117-12-4 EINECS: 418-140-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...