(Propargyloxy)trimethylsilane 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Trimethyl(prop-2-yn-1-yloxy)silane
L12936.06
L12936.14
L12936.06EA
1170
CZK
L12936.06
L12936.14
(Propargyloxy)trimethylsilane 97%
(Propargyloxy)trimethylsilane
Purity: 97%
CAS No.: 5582-62-7
Molecular Formula: C6H12OSi
Formula Weight: 128.25
Storage Temperature: 2°C to 8°C
Flash Point: 8°(46°F)
Refractive Index: 1.4080
Boiling Point: 110-111°
Beilstein Reference: 2036793
UN No.: UN1993
MDL No.: MFCD00042844
Vzorec:
C₆H₁₂OSi MW: 128.25 g/mol Bod varu: 110…111 °C Hustota: 0.833 Bod vzplanutí: 8 °C (46 °F) |
MDL Number:
MFCD00042844 CAS číslo: 5582-62-7 EINECS: 226-981-7 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...