4-n-Butoxyphenyl isocyanate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L12877.03
L12877.06
L12877.03EA
1030
CZK
L12877.03
L12877.06
4-n-Butoxyphenyl isocyanate ≥98%
4-n-Butoxyphenyl isocyanate
Purity: 98%
CAS No.: 28439-86-3
Molecular Formula: C11H13NO2
Formula Weight: 191.23
Storage Temperature: 2°C to 8°C
Flash Point:
Vzorec:
C₁₁H₁₃NO₂ Bod varu: 149…150 °C (3 mmHg) Hustota: 1,055 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00037042 CAS číslo: 28439-86-3 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...