Tisk…

5-Chlorobenzo[b]thiophene-3-acetic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

L12836.03 L12836.MD
L12836.03EA 3230 CZK
L12836.03 L12836.MD
5-Chlorobenzo[b]thiophene-3-acetic acid 97%
5-Chlorobenzo[b]thiophene-3-acetic acid

Purity: 97% CAS No.: 17266-30-7 Molecular Formula: C10H7ClO2S Formula Weight: 226.68 Melting Point: 152-155° Beilstein Reference: 1244795 MDL No.: MFCD00052308

Vzorec: C₁₀H₇ClO₂S
Bod tání: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00052308
CAS číslo: 17266-30-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR