Isobutyl acrylate ≥99% stabilizovaný
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 100 ppm
stab with 100ppm 4-methoxyphenol
Purity: 99%
CAS No.: 106-63-8
Molecular Formula: C7H12O2
Formula Weight: 128.17
Flash Point: 32°(89°F)
Refractive Index: 1.4150
Melting Point: -61°
Boiling Point: 139°
Beilstein Reference: 1749388
UN No.: UN2527
MDL No.: MFCD00042865
Vzorec:
C₇H₁₂O₂ Bod varu: 139 °C Bod tání: –61 °C Hustota: 0,888 Bod vzplanutí: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042865 CAS číslo: 106-63-8 EINECS: 203-417-8 UN: 2527 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...