Tisk…

Triethylvinylsilane 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Vinyltriethylsilane

L12461.03 L12461.06
L12461.03EA 1020 CZK
L12461.03 L12461.06
Triethylvinylsilane 97%
Triethylvinylsilane

Purity: 97% CAS No.: 1112-54-5 Molecular Formula: C8H18Si Formula Weight: 142.32 Flash Point: 25°(77°F) Refractive Index: 1.4340 Boiling Point: 145-146° Beilstein Reference: 1743830 UN No.: UN1993 MDL No.: MFCD00041345

Vzorec: C₈H₁₈Si
MW: 142,32 g/mol
Bod varu: 145…146 °C
Hustota: 0,771
Bod vzplanutí: 25 °C (77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041345
CAS číslo: 1112-54-5
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR