Tisk…

1-Dimethylamino-2-propyne 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: N,N-Dimethylpropargylamine, N,N-Dimethyl-2-propyn-1-amine

L12336.14 L12336.06
L12336.14EA 4090 CZK
L12336.14 L12336.06
1-Dimethylamino-2-propyne 98%
1-Dimethylamino-2-propyne

Purity: 98% CAS No.: 7223-38-3 Molecular Formula: HC≡CCH2N(CH3)2 Formula Weight: 83.13 Flash Point: -7°(19°F) Refractive Index: 1.4200 Boiling Point: 80-81° Beilstein Reference: 1071222 UN No.: UN2733 MDL No.: MFCD00008575

Vzorec: C₅H₉N
MW: 83,13 g/mol
Bod varu: 80…81 °C
Hustota: 0,772
Bod vzplanutí: –7 °C (19 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008575
CAS číslo: 7223-38-3
EINECS: 230-620-9
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR