Tisk…

Malonic acid dihydrazide 99%

Dodavatel: Thermo Scientific
Varování

Synonyms: Malonohydrazide

L12270.06 L12270.14
L12270.06EA 2400 CZK
L12270.06 L12270.14
Malonic acid dihydrazide 99%
Malonic acid dihydrazide

Purity: 99% CAS No.: 3815-86-9 Molecular Formula: C3H8N4O2 Formula Weight: 132.12 Melting Point: 152-156° Beilstein Reference: 1072337 MDL No.: MFCD00041268

Vzorec: C₃H₈N₄O₂
MW: 132,12 g/mol
Bod tání: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00041268
CAS číslo: 3815-86-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR