2-Hexyldecanoic acid 97%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Isopalmitic Acid, Pentadecane-7-carboxylic Acid
L12238.14
L12238.22
L12238.14EA
1030
CZK
L12238.14
L12238.22
2-Hexyldecanoic acid 97%
2-Hexyldecanoic acid
Purity: 97%
CAS No.: 25354-97-6
Molecular Formula: C16H32O2
Formula Weight: 256.43
Flash Point: 174°(345°F)
Refractive Index: 1.4460
Melting Point: 10°
Boiling Point: 165-168°/2mm
Beilstein Reference: 1777217
MDL No.: MFCD00059651
Vzorec:
C₁₆H₃₂O₂ MW: 256.43 g/mol Bod varu: 165…168 °C (2 mmHg) Hustota: 0.880 Bod vzplanutí: 174 °C (345 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00059651 CAS číslo: 25354-97-6 EINECS: 246-885-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...