Tisk…

3-Diethylamino-1-propyne ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: N,N-Diethylpropargylamine

L12237.06 L12237.14
L12237.06EA 1840 CZK
L12237.06 L12237.14
3-Diethylamino-1-propyne ≥98%
3-Diethylamino-1-propyne

Purity: 98+% CAS No.: 4079-68-9 Molecular Formula: C7H13N Formula Weight: 111.19 Flash Point: 20°(68°F) Refractive Index: 1.4305 Odour: Amine-like Boiling Point: 119-120° Beilstein Reference: 1098468 UN No.: UN2733 MDL No.: MFCD00039881

Vzorec: C₇H₁₃N
MW: 111,19 g/mol
Bod varu: 119…120 °C
Hustota: 0,804
Bod vzplanutí: 20 °C (68 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00039881
CAS číslo: 4079-68-9
EINECS: 223-804-5
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR