Tisk…

Bis(2-benzamidophenyl)disulphide 97%

Dodavatel: Thermo Scientific

Synonyms: 2,2'-Dithiodibenzanilide, 2,2'-Dibenzamidodiphenyl Disulphide, Bis(2-benzamidophenyl) Disulphide

L12110.14 L12110.22
L12110.14EA 1140 CZK
L12110.14 L12110.22
Bis(2-benzamidophenyl)disulphide 97%
Bis(2-benzamidophenyl)disulphide

Purity: 97% CAS No.: 135-57-9 Molecular Formula: C26H20N2O2S2 Formula Weight: 456.59 Melting Point: 140-143° Beilstein Reference: 721443 MDL No.: MFCD00043806

Vzorec: C₂₆H₂₀N₂O₂S₂
MW: 456,58 g/mol
Bod tání: 140…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00043806
CAS číslo: 135-57-9
EINECS: 205-201-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR