1-Adamantaneacetic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-(Adamantan-1-yl)acetic acid, 1-Carboxymethyladamantane
L12029.03
L12029.06
L12029.14
L12029.03EA
930
CZK
L12029.03
L12029.06
L12029.14
1-Adamantaneacetic acid ≥98%
1-Adamantaneacetic acid
Purity: 98+%
CAS No.: 4942-47-6
Molecular Formula: C12H18O2
Formula Weight: 194.28
Melting Point: 136-138°
Beilstein Reference: 641412
MDL No.: MFCD00074728
Vzorec:
C₁₂H₁₈O₂ MW: 194,27 g/mol Bod tání: 136…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074728 CAS číslo: 4942-47-6 EINECS: 225-585-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...