Tisk…

1-Isocyanato-3-(methylthio)benzene 96%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 3-(Methylthio)phenyl isocyanate

L12008.03 L12008.06
L12008.03EA 1480 CZK
L12008.03 L12008.06
1-Isocyanato-3-(methylthio)benzene 96%
1-Isocyanato-3-(methylthio)benzene

Purity: 96% CAS No.: 28479-19-8 Molecular Formula: C8H7NOS Formula Weight: 165.22 Storage Temperature: 2°C to 8°C Flash Point: 113°(235°F) Refractive Index: 1.6030 Boiling Point: 129-130°/5mm Beilstein Reference: 387507 UN No.: UN2206 MDL No.: MFCD00013863

Vzorec: C₈H₇NOS
MW: 165,22 g/mol
Bod varu: 129…130 °C (5 mmHg)
Hustota: 1,199
Bod vzplanutí: 113 °C (235 °F)
MDL Number: MFCD00013863
CAS číslo: 28479-19-8
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR