Dimethyl-trans-stilbene-4,4'-dicarboxylate ≥98%
Dodavatel: Thermo Fisher Scientific
L11969.03
L11969.MD
L11969.03EA
4190
CZK
L11969.03
L11969.MD
Dimethyl-trans-stilbene-4,4'-dicarboxylate ≥98%
Dimethyl-trans-stilbene-4,4'-dicarboxylate
Purity: 98+%
CAS No.: 34541-73-6
Molecular Formula: C18H16O4
Formula Weight: 296.33
Melting Point: 234-236°
Beilstein Reference: 2471189
MDL No.: MFCD00039527
Vzorec:
C₁₈H₁₆O₄ Bod tání: 234…236 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039527 CAS číslo: 34541-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...