Tisk…

Sodium-4-aminoazobenzene-4'-sulphonate 90%, tech.

Dodavatel: Thermo Fisher Scientific
Varování

L11965.14 L11965.22
L11965.14EA 1340 CZK
L11965.14 L11965.22
Sodium-4-aminoazobenzene-4'-sulphonate 90%, tech.
Sodium-4-aminoazobenzene-4'-sulphonate

Purity: 90% CAS No.: 2491-71-6 Molecular Formula: C12H10N3NaO3S Formula Weight: 299.29 Beilstein Reference: 8098548 MDL No.: MFCD00035564

Vzorec: C₁₂H₁₁N₃O₃S
Storage Temperature: Ambient
MDL Number: MFCD00035564
CAS číslo: 2491-71-6
EINECS: 219-657-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR