Tisk…

N-(2-Hydroxypropyl)morpholine 98%

Dodavatel: Thermo Fisher Scientific
Varování

L11859.09 L11859.18
L11859.09EA 1800 CZK
L11859.09 L11859.18
N-(2-Hydroxypropyl)morpholine 98%
N-(2-Hydroxypropyl)morpholine

Purity: 98% CAS No.: 2109-66-2 Molecular Formula: C7H15NO2 Formula Weight: 145.20 Flash Point: 92°(197°F) Refractive Index: 1.4635 Boiling Point: 218° Beilstein Reference: 1443 MDL No.: MFCD00023378

Vzorec: C₇H₁₅NO₂
Bod varu: 218 °C
Hustota: 1,020
Bod vzplanutí: 92 °C (197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00023378
CAS číslo: 2109-66-2
EINECS: 218-298-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR