5-Chloro-2,4-dinitrotoluene 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-Chloro-5-methyl-2,4-dinitrobenzene
L11808.06
L11808.14
L11808.06EA
2150
CZK
L11808.06
L11808.14
5-Chloro-2,4-dinitrotoluene 97%
5-Chloro-2,4-dinitrotoluene
Purity: 97%
CAS No.: 51676-74-5
Molecular Formula: C7H5ClN2O4
Formula Weight: 216.58
Melting Point: 89-91°
Beilstein Reference: 2506591
UN No.: UN2811
MDL No.: MFCD00041235
Vzorec:
C₇H₅ClN₂O₄ MW: 216,58 g/mol Bod tání: 89…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041235 CAS číslo: 51676-74-5 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...