Tisk…

Sodium-p-styrenesulphonate hydrate 90%, tech.

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: p-Styrenesulphonic acid sodium salt hydrate, Sodium-p-styrenesulphonate hydrate, Sodium 4-vinylbenzenesulphonate hydrate, Styrene-4-sulphonic acid sodium salt hydrate

L11685.06 L11685.14
L11685.06EA 821 CZK
L11685.06 L11685.14
Sodium-p-styrenesulphonate hydrate 90%, tech.
Sodium-p-styrenesulphonate hydrate

Purity: 90% CAS No.: 2695-37-6 Molecular Formula: C8H7NaO3S.xH2O Formula Weight: 206.20(anhy) Melting Point: 320° Beilstein Reference: 3575931 MDL No.: MFCD03092905

Vzorec: C₈H₇NaO₃S·xH₂O
MW: 206.19 g/mol (anhydre)
Bod tání: >320 °C
Storage Temperature: Ambient
MDL Number: MFCD03092905
CAS číslo: 2695-37-6
EINECS: 220-266-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR